Would you like to log in again?
SCIENCEWARE® Filtering Flask with Side Arm, Bel-Art
Supplier: Bel-Art Products
H38941-0000
47750-576EA 47.87 USD InStock
47750-576
SCIENCEWARE® Filtering Flask with Side Arm, Bel-Art
Flasks Filter Flasks
Polypropylene plastic flask has side-arm at 45˚ downward angle to prevent tipping.
  • Polypropylene construction
  • Downward angle prevents tiping
  • Flexible side arm
Arm accepts flexible tubing from ⅜" to ½" I.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty