Value Laboratory Refrigerators, Thermo Scientific
05LREETSA 10LCEETSA
10829-220EA 2350.08 USD
10829-220 89006-900
Value Laboratory Refrigerators, Thermo Scientific
Value laboratory refrigerators are an economical solution for safe sample storage.
  • NRTL listed and factory tested prior to shipment
  • Adjustable temperature control
  • Environmentally friendly non-CFC refrigerants
  • Easy-to-clean, durable finish
  • Not suitable for corrosive environments
These units are intended for basic laboratory use only. Storage of high-value life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features.
Certifications: CSA approved. UL listed.