"Electro-Flo" Shut-Off Valve, Ace Glass Incorporated
12165-26 12165-48 12165-14 12165-57 12165-20 12165-54
89076-702EA 498.55 USD
89076-702 89076-700 89076-698 89076-696 89076-694 89076-692
"Electro-Flo" Shut-Off Valve, Ace Glass Incorporated
A water or air shut-off solenoid valve for use with water-flo power cut-off or as a general laboratory shut-off valve
A pre-filter is recommended when this product is used with water or air, 15 micron or less.
Operates from 0.35Kg/cm2 to 10.5Kg/cm2 (5 to 150 psi) for water or 0.35Kg/cm2 for air and up to 91°C. Internal design of pilot-operated, piston-type, valve assures exceptional flow performance. Constructed of cast bronze with waterproof cast-coil that has a lifetime warranty. Valve must be installed in horizontal piping with solenoid in vertical position. Supplied with 1.8 meter grounded cord and female pipe thread.