Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
New Brunswick Premium U570 ULT Upright Freezer, Eppendorf
U9270-0000 U9270-0002 U9270-0002 U9270-0000
97048-916EA 19623.98 USD InStock
97048-916 CA97048-918 97048-918 CA97048-916
New Brunswick Premium U570 ULT Upright Freezer, Eppendorf
Freezers Laboratory / General Purpose Freezers
The U570 is the second largest upright freezer in the Premium lineup
The U570 will provide years of maintenance-free, reliable operation with minimal energy consumption. The U570 consumes 14.1 kWh/day and pulls down from ambient to set temperature in 5.3 hours, roughly twice as fast compared to similar sized competitive models. A bright eye level, LED control system is standard along with a 5 compartment, adjustable shelving system which can accommodate any type of racking configuration
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty