Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
New Brunswick™ Galaxy® 170 R High-Capacity CO₂ Incubators, Eppendorf
Supplier: Eppendorf
CO17231005 CO17214005 CO170R1200200 CO17214065 CO17224005 CO17211005 CO170R1200000
75875-218EA 13037.33 USD InStock
75875-218 75875-212 89171-954 75875-214 75875-216 89171-956 89171-952
New Brunswick™ Galaxy® 170 R High-Capacity CO₂ Incubators, Eppendorf
Incubators CO₂ Incubators
Galaxy® 170 R high-capacity CO2 incubators can be conveniently placed on or under the bench or even double-stacked, making them ideal for a wide range of specialized applications. The 170 R is the advanced model of the 170 series.
  • Sealed inner glass door
  • Reliable IR CO2 sensor
  • RS-232 communication port
  • 25 mm access ports
  • Low gas consumption
  • Alarm relay standard
The Galaxy® 170 R CO2 Incubators are high-capacity, 170 liter incubators that provide superior quality, outstanding performance, and intuitive operation
Fanless design offers more usable space and eliminates the fan, an often repeated source of contamination. A six-sided direct heating system provides a uniform incubated environment to gently bathe cells, and the deep-drawn, stainless steel chamber eliminates seams or welds and allows easy cleaning. Features an LCD display with display readout of temperature and CO2 level, 72-hour data/chart logger, and alarm settings.
Galaxy® 170 R models provide a tremendous amount of usable space that can be optimized with options such as high temperature disinfection (4 hour at 120® °C), two O2 control options (0.1–19%, 1–19%) for hypoxic studies, split inner doors, copper chamber and interior for optimized protection against contamination, and humidity alert package.
For all units, CO2 control is ±0.1%, CO2 range is 0.2–20%, CO2 stability at 5% CO2 is ±0.2%, and CO2 uniformity is ±0.1%.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty