Calcium stearate USP

Supplier: ACROS ORGANICS MS

AC61243-0030
200024-660EA 168.26 USD InStock
200024-660
Calcium stearate USP
Calcium stearate
Formula: C₃₆H₇₀O₄·Ca
MW: 607,03 g/mol
CAS Number: 66071-81-6

Order Now

Specification Test Results

Parameter EINECS 266-106-6
Molecular Weight 607.03

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR