VWR® Heat-Resistant Films for Real-Time qPCR, Ultra-Clear Polyester, Raised Rim
Supplier: VWR International
VWR®
TSSRT2RVWR100
89024-752CS 194.91 USD InStock
89024-752
VWR® Heat-Resistant Films for Real-Time qPCR, Ultra-Clear Polyester, Raised Rim
Seals Microplate Sealing Films and Foils
Heat-resistant films feature strong, non-absorbing, non-fluorescing medical-grade adhesive for superior performance in real-time qPCR applications and for sealing PCR plates.
  • Transparent films
  • Raised rim
  • Nonsterile
Transparent polyester films exhibit ultra-high optical clarity
Ideal for real-time PCR applications requiring maximum optical clarity.
The 125µm film resists wrinkling in applications involving freezing. All films are supplied RNase-, DNase-, and nuclease-free. Sized to fit within within the edges of a raised-rim plate. Recommended for use in temperatures from -40 to 120°C (-40 to 248°F).