Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
SOURCE 15RPC ST Columns, GE Healthcare
Supplier: GE Healthcare - Life Sciences
SOURCE™
17-5068-01 17-5068-01 17-5068-01
89125-750EA 960.22 USD InStock
89125-750 CA95017-164 95017-164
SOURCE 15RPC ST Columns, GE Healthcare
Chromatography Columns
Source 15RPC ST 4.6/100 is a prepacked stainless steel column which provides fast, high-capacity preparative protein purification with reversed phase chromatography.
  • Wide pH range (1-12), outstanding selectivity, chemical resistance, high capacity, and high resolution at high flow rates
  • Excellent scalability, from RESOURCE to FineLINE columns
  • Using ÄKTA design and other high-performance liquid chromatography systems
SOURCE 15RPC is based on rigid, monosized 15 µm diameter polystyrene/divinyl benzene beads. The matrix has outstanding selectivity for RPC. The monodispersity of the beads yields stable beds, low back pressures and excellent results at high flow rates.With its high physical and chemical stability and very high batch-to-batch reproducibility, SOURCE 15RPC is well suited for all stages of an industrial scale operation - from research and process development through scale-up and into production. SOURCE 15RPC offers properties superior to those of other polymeric matrices.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty