Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
MAL-dPEG®4-(m-dPEG®11)3
Supplier: QUANTA BIODESIGN MS
11406-1000MG 11406-100MG
102856-568EA 1700 USD InStock
102856-568 102856-566
MAL-dPEG®4-(m-dPEG®11)3
MAL-dPEG®4-(m-dPEG®11)3
Single compound - dPEG® Spacer is 64 atoms and 75.8 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
https://us.vwr.com/stibo/web/std.lang.all/63/76/12446376.jpg
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty