Amino-dPEG®11-ONH-t-boc

Synonyms: Amino-dPEG®11-ONH-t-boc

11112-1000MG 11112-100MG
102856-552EA 1647.69 USD
102856-552 102856-550
Amino-dPEG®11-ONH-t-boc
Amino-PEG11-ONH-t-boc

Single compound - dPEG® Spacer is 38 atoms and 43.8 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₉H₆₀N₂O₁₄
MW: 660.79 g/mol
CAS Number: 1630094-83-5
https://us.vwr.com/stibo/web/std.lang.all/63/68/12446368.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR