Biotin-dPEG®11-oxyamine.HCl
11102-1000MG 11102-50MG 11102-100MG
102856-540EA 481.23 USD
102856-540 102856-542 102856-538
Biotin-dPEG®11-oxyamine.HCl
Biotin-PEG11-oxyamine.HCl
Single compound - dPEG® Spacer is 38 atoms and 44.9 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
https://us.vwr.com/stibo/web/std.lang.all/63/19/12446319.jpg