Fmoc-N-amido-dPEG®12-NHS ester

Synonyms: Fmoc-N-amido-dPEG®12-NHS ester

10996-100MG 10996-1000MG
102856-530EA 253.69 USD
102856-530 102856-532
Fmoc-N-amido-dPEG®12-NHS ester
Fmoc-N-amido-PEG12-NHS ester

Single compound - dPEG® Spacer is 40 atoms and 46.5 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₆H₆₈N₂O₁₈
MW: 937.03 g/mol
CAS Number: 488085-18-3
https://us.vwr.com/stibo/web/std.lang.all/63/15/12446315.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR