Fmoc-N-amido-dPEG®4-NHS ester

10994-100MG 10994-1000MG
102856-522EA 171.31 USD
102856-522 102856-524
Fmoc-N-amido-dPEG®4-NHS ester
Fmoc-N-amido-dPEG4-NHS ester

Single compound - dPEG® Spacer is 17 atoms and 18.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₀H₃₆N₂O₁₀
MW: 584.24 g/mol
CAS Number: 1314378-14-7
https://us.vwr.com/stibo/web/std.lang.all/63/11/12446311.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR