Bis-dPEG®17-PFP ester

10980-100MG 10980-1000MG
102856-492EA 1313.25 USD
102856-492 102856-490
Bis-dPEG®17-PFP ester
Bis-PEG17-PFP ester

Single compound - dPEG® Spacer is 55 atoms and 64.4 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₅₀H₇₂F₁₀O₂₁
MW: 1199.078 g/mol
MDL Number: MFCD21363294
CAS Number: 1334170-00-1
https://us.vwr.com/stibo/web/std.lang.all/19/22/12381922.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR