Bis-dPEG®25-NHS ester

10968-1000MG 10968-100MG
102856-478EA 481.23 USD
102856-478 102856-480
Bis-dPEG®25-NHS ester
Bis-PEG25-NHS ester

Single compound - dPEG® Spacer is 79 atoms and 93.0 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₆₂H₁₁₂N₂O₃₃
MW: 1413.546 g/mol
MDL Number: MFCD21363290
CAS Number: 1008402-79-6
https://us.vwr.com/stibo/web/std.lang.all/19/16/12381916.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR