Amino-dPEG®12-t-boc-hydrazide

Synonyms: Amino-dPEG®12-t-boc-hydrazide

10958-100MG 10958-1000MG
102856-464EA 1442.38 USD
102856-464 102856-462
Amino-dPEG®12-t-boc-hydrazide
Amino-PEG12-t-boc-hydrazide

Single compound - dPEG® Spacer is 45 atoms and 50.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₂H₆₅N₃O₁₅
MW: 731.87 g/mol
CAS Number: 1334169-97-9
https://us.vwr.com/stibo/web/std.lang.all/19/08/12381908.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR