Bis-dPEG®21-NHS ester
Single compound - dPEG® Spacer is 67 atoms and 79.1 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
Formula:
C₅₄H₉₆N₂O₂₉ MW: 1237.34 g/mol |
CAS Number:
1008402-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...