Bis-dPEG®13-NHS ester
Single compound - dPEG® Spacer is 43 atoms and 50.1 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
Formula:
C₃₈H₆₄N₂O₂₁ MW: 884.916 g/mol |
MDL Number:
MFCD21363287 CAS Number: 1008402-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...