m-dPEG®23-alcohol

10942-100MG 10942-1000MG
102856-440EA 893.15 USD
102856-440 102856-438
m-dPEG®23-alcohol
m-PEG23-alcohol

Single compound - dPEG® Spacer is 70 atoms and 83.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₇H₉₆O₂₄
MW: 1045.251 g/mol
MDL Number: MFCD21363223
CAS Number: 114740-40-8
https://us.vwr.com/stibo/web/std.lang.all/18/96/12381896.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR