m-dPEG®48-MAL

Synonyms: Methyl-PEG48-Maleimidem-PEG48-Maleimide

10932-100MG 10932-1000MG
102856-424EA 2059.62 USD
102856-424 102856-422
m-dPEG®48-MAL
m-PEG48-Mal

Single compound - dPEG® Spacer is 152 atoms and 178.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₀₄H₂₀₂N₂O₅₁
MW: 2296.7 g/mol
CAS Number: 88504-24-9
https://us.vwr.com/stibo/web/std.lang.all/18/92/12381892.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR