Amino-dPEG®36-acid

10907-100MG 10907-1000MG
102856-354EA 411.92 USD
102856-354 102856-356
Amino-dPEG®36-acid
Amino-PEG36-acid

Single compound - dPEG® Spacer is 111 atoms and 132.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₇₅H₁₅₁NO₃₈
MW: 1674.985 g/mol
MDL Number: MFCD21363230
CAS Number: 663921-15-1
https://us.vwr.com/stibo/web/std.lang.all/10/01/12381001.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR