QUBD10849
10849-100MG 10849-1000MG
102856-244EA 180.25 USD
102856-244 102856-246
QUBD10849
Single compound - dPEG® Spacer is 41 atoms and 47.6 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.