Biotin-dPEG®7-NH2

Synonyms: Biotin-dPEG®7-NH2

10826-100MG 10826-1000MG
102856-230EA 1235.77 USD
102856-230 102856-228
Biotin-dPEG®7-NH2
Biotin-PEG7-NH2

Single compound - dPEG® Spacer is 25 atoms and 29.8 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₆H₅₀N₄O₉S
MW: 594.76 g/mol
CAS Number: 1334172-76-7
https://us.vwr.com/stibo/web/std.lang.all/09/60/12380960.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR