MAL-dPEG®3-lipoamido

Synonyms: MAL-dPEG®3-lipoamido

10817-1000MG 10817-100MG
102856-210EA 1235.77 USD
102856-210 102856-208
MAL-dPEG®3-lipoamido
MAL-PEG3-lipoamido

Single compound - dPEG® Spacer is 27 atoms and 31.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₅H₄₁N₃O₇S₂
MW: 559.74 g/mol
CAS Number: 1334172-72-3
https://us.vwr.com/stibo/web/std.lang.all/09/50/12380950.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR