Lipoamido-dPEG®12-acid

10808-100MG 10808-1000MG
102856-198EA 1717 USD
102856-198 102856-196
Lipoamido-dPEG®12-acid
Lipoamido-PEG12-acid

Single compound - dPEG® Spacer is 48 atoms and 55.5 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₅H₆₇NO₁₅S₂
MW: 806.034 g/mol
MDL Number: MFCD21363244
CAS Number: 1334172-71-2
https://us.vwr.com/stibo/web/std.lang.all/09/46/12380946.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR