Lipoamido-dPEG®8-acid

10807-100MG 10807-1000MG
102856-192EA 343.92 USD
102856-192 102856-194
Lipoamido-dPEG®8-acid
Lipoamido-PEG8-acid

Single compound - dPEG® Spacer is 36 atoms and 41.2 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₇H₅₁NO₁₁S₂
MW: MFCD21363243
CAS Number: 1334172-70-1
https://us.vwr.com/stibo/web/std.lang.all/09/44/12380944.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR