Biotin-dPEG®23-azide

10787-100MG 10787-1000MG
102856-156EA 411.92 USD
102856-156 102856-158
Biotin-dPEG®23-azide
Biotin-PEG23-azide

Single compound - dPEG® Spacer is 73 atoms and 87.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₅₈H₁₁₂N₆O₂₅S
MW: 1325.6 g/mol
MDL Number: MFCD21363314
CAS Number: 956494-20-5
https://us.vwr.com/stibo/web/std.lang.all/09/26/12380926.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR