NHS-dPEG®24-biotin

10774-1000MG
102856-136EA 2013.85 USD
102856-136
NHS-dPEG®24-biotin
NHS-PEG24-biotin

Single compound - dPEG® Spacer is 81 atoms and 97.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₆₅H₁₂₀N₄O₃₀S
MW: 1469.72 g/mol
CAS Number: 365441-71-0
https://us.vwr.com/stibo/web/std.lang.all/09/15/12380915.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR