m-dPEG®4-MAL

Synonyms: m-PEG4-Maleimidemethyl-PEG4-Maleimide

10745-100MG 10745-1000MG
102856-100EA 618.54 USD
102856-100 102856-098
m-dPEG®4-MAL
m-PEG4-Mal

Single compound - dPEG® Spacer is 20 atoms and 22.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₆H₂₆N₂O₇
MW: 360.4 g/mol
CAS Number: 1263044-81-0
https://us.vwr.com/stibo/web/std.lang.all/08/97/12380897.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR