Bis-dPEG®3-NHS ester

Synonyms: Bis-dPEG®3-NHS ester

10726-100MG 10726-1000MG
102856-088EA 460.31 USD
102856-088 102856-086
Bis-dPEG®3-NHS ester
Bis-PEG3-NHS ester

Single compound - dPEG® Spacer is 13 atoms and 14.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₈H₂₄N₂O₁₁
MW: 444.39 g/mol
CAS Number: 1314378-16-9
https://us.vwr.com/stibo/web/std.lang.all/08/93/12380893.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR