Fmoc-N-Lys-(dPEG®4-biotin)-OH-(acid)

Synonyms: Fmoc-N-Lys-(dPEG®4-biotin)-OH-(acid)

10613-100MG 10613-1000MG
102856-032EA 1373.08 USD
102856-032 102856-030
Fmoc-N-Lys-(dPEG®4-biotin)-OH-(acid)
Fmoc-N-Lys-(PEG4-biotin)-OH-(acid)

Single compound - dPEG® Spacer is 19.1 atoms and 16 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₂H₅₉N₅O₁₁S
MW: 842.01 g/mol
CAS Number: 1334172-64-3
https://us.vwr.com/stibo/web/std.lang.all/08/67/12380867.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR