Azido-dPEG®36-alcohol

10544-100MG 10544-1000MG
102856-022EA 1305.08 USD
102856-022 102856-020
Azido-dPEG®36-alcohol
Azido-PEG36-alcohol

Single compound - dPEG® Spacer is 108 atoms and 129.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₇₂H₁₄₅N₃O₃₆
MW: 1628.92 g/mol
MDL Number: MFCD21363269
CAS Number: 73342-16-2
https://us.vwr.com/stibo/web/std.lang.all/08/62/12380862.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR