Azido-dPEG®24-alcohol

10543-100MG 10543-1000MG
102856-016EA 468.15 USD
102856-016 102856-018
Azido-dPEG®24-alcohol
Azido-PEG24-alcohol

Single compound - dPEG® Spacer is 72 atoms and 86.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₈H₉₇N₃O₂₄
MW: 1100.29 g/mol
CAS Number: 73342-16-2
https://us.vwr.com/stibo/web/std.lang.all/08/60/12380860.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR