Azido-dPEG®4-alcohol

Synonyms: PEG4-Azide11-Azido-3,6,9-trioxaundecanol

10541-100MG 10541-1000MG
102856-010EA 206.62 USD
102856-010 102856-008
Azido-dPEG®4-alcohol
11-Azido-3,6,9-trioxaundecanol

Single compound - dPEG® Spacer is 14 atoms and 15.3 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₈H₁₇N₃O₄
MW: 219.24 g/mol
MDL Number: MFCD03701128
CAS Number: 86770-67-4
https://us.vwr.com/stibo/web/std.lang.all/08/55/12380855.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR