m-dPEG®8-azide (azido-m-dPEG®8)

10534-1000MG 10534-100MG
102855-996EA 240.62 USD
102855-996 102855-998
m-dPEG®8-azide (azido-m-dPEG®8)
m-PEG8-azide (azido-m-PEG8)

Single compound - dPEG® Spacer is 26 atoms and 29.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₇H₃₅N₃O₈
MW: 409.48 g/mol
CAS Number: 869718-80-9
https://us.vwr.com/stibo/web/std.lang.all/08/49/12380849.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR