Azido-dPEG®23-amine

10525-1000MG 10525-100MG
102855-980EA 411.92 USD
102855-980 102855-982
Azido-dPEG®23-amine
Azido-PEG23-amine

Single compound - dPEG® Spacer is 72 atoms and 86.8 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₈H₉₈N₄O₂₃
MW: 1099.3 g/mol
CAS Number: 749244-38-0
https://us.vwr.com/stibo/web/std.lang.all/08/41/12380841.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR