Azido-dPEG®7-amine
Synonyms:
Azido-dPEG®7-amine
Single compound - dPEG® Spacer is 25 atoms and 28.8 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
Formula:
C₁₆H₃₄N₄O₇ MW: 394.46 g/mol |
CAS Number:
1333154-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...