Azido-dPEG®12-NHS ester

Synonyms: Azido-dPEG®12-NHS ester

10505-100MG 10505-1000MG
102855-948EA 1030.46 USD
102855-948 102855-946
Azido-dPEG®12-NHS ester
Azido-PEG12-NHS ester

Single compound - dPEG® Spacer is 28 atoms and 32.2 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₁H₅₆N₄O₁₆
MW: 740.794 g/mol
MDL Number: MFCD13184947
CAS Number: 1108750-59-9
https://us.vwr.com/stibo/web/std.lang.all/08/24/12380824.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR