Azido-dPEG®4-NHS ester

Synonyms: N-Succinimidyl 15-Azido-4,7,10,13-tetraoxapentadecanoateNHS-PEG4-Azide15-Azido-4,7,10,13-tetraoxapentadecanoic acid N-succinimidyl ester

10501-100MG 10501-1000MG
102855-936EA 686.54 USD
102855-936 102855-934
Azido-dPEG®4-NHS ester
Azido-PEG4-NHS ester

Single compound - dPEG® Spacer is 16 atoms and 17.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₅H₂₄N₄O₈
MW: 388.38 g/mol
MDL Number: MFCD13184948
CAS Number: 944251-24-5
https://us.vwr.com/stibo/web/std.lang.all/08/18/12380818.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR