QUBD10424
10424-100MG 10424-1000MG
102855-908EA 927 USD
102855-908 102855-906
QUBD10424
Single compound - dPEG® Spacer is 38 atoms and 86.4 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.