MAL-dPEG®4-(m-dPEG®4)3

Synonyms: MAL-dPEG®4-(m-dPEG®4)3

10416-1000MG 10416-100MG
102855-892EA 1785 USD
102855-892 102855-890
MAL-dPEG®4-(m-dPEG®4)3
MAL-PEG4-(m-PEG4)3

Single compound - dPEG® Spacer is 43 atoms and 47.9 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₅₈H₁₀₆N₆O₂₆
MW: 1303.49 g/mol
CAS Number: 1333154-72-5
https://us.vwr.com/stibo/web/std.lang.all/07/96/12380796.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR