DNP-dPEG®12-NHS ester

10399-1000MG 10399-100MG
102855-862EA 343.92 USD
102855-862 102855-864
DNP-dPEG®12-NHS ester
DNP-PEG12-NHS ester

Single compound - dPEG® Spacer is 40 atoms and 46.4 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₇H₆₀N₄O₂₀
MW: 880.887 g/mol
MDL Number: MFCD13185013
CAS Number: 1334178-01-6
https://us.vwr.com/stibo/web/std.lang.all/07/81/12380781.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR