QUBD10364
10364-1000MG 10364-100MG
102855-804EA 1236 USD
102855-804 102855-802
QUBD10364
Single compound - dPEG® Spacer is 74 atoms and 86.4 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.