QUBD10363
10363-1000MG 10363-100MG
102855-798EA 103 USD
102855-798 102855-800
QUBD10363
Single compound - dPEG® Spacer is 27 atoms and 29.8 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.