QUBD10362
10362-100MG 10362-1000MG
102855-796EA 618 USD
102855-796 102855-794
QUBD10362
Single compound - dPEG® Spacer is 14 atoms and 16.5 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.