m-dPEG®24-MAL

Synonyms: m-PEG24-MaleimideMethyl-PEG24-Maleimide

10319-100MG 10319-1000MG
102855-690EA 396.23 USD
102855-690 102855-692
m-dPEG®24-MAL
m-PEG24-Mal

Single compound - dPEG® Spacer is 80 atoms and 50.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₅₆H₁₀₆N₂O₂₇
MW: 1239.45 g/mol
MDL Number: MFCD11041152
CAS Number: 88504-24-9
https://us.vwr.com/stibo/web/std.lang.all/07/00/12380700.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR