m-dPEG®24-amine

10318-1000MG 10318-100MG
102855-686EA 273.31 USD
102855-686 102855-688
m-dPEG®24-amine
m-PEG24-amine

Single compound - dPEG® Spacer is 74 atoms and 86.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₉H₁₀₁NO₂₄
MW: 1088.319 g/mol
MDL Number: MFCD11041163
CAS Number: 32130-27-1
https://us.vwr.com/stibo/web/std.lang.all/06/97/12380697.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR