Fmoc-N-amido-dPEG®24-acid
Single compound - dPEG® Spacer is 76 atoms and 89 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
Formula:
C₆₆H₁₁₃NO₂₈ MW: 1368,59 g/mol |
CAS Number:
756526-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...