Amino-dPEG®12-t-butyl ester
Single compound - dPEG® Spacer is 76 atoms and 89 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
Formula:
C₅₅H₁₁₁NO₂₆ MW: 1202.46 g/mol |
MDL Number:
MFCD11041158 CAS Number: 872340-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...