m-dPEG®24-NHS ester

10304-1000MG 10304-100MG
102855-646EA 231.75 USD
102855-646 102855-648
m-dPEG®24-NHS ester
m-PEG24-NHS ester

Single compound - dPEG® Spacer is 74 atoms and 86.2 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₅₄H₁₀₃NO₂₈
MW: 1214.386 g/mol
MDL Number: MFCD11041155
CAS Number: 756525-94-7
https://us.vwr.com/stibo/web/std.lang.all/06/76/12380676.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR